missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LIPI Full-length ORF (AAI48692.1, 1 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00149998-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MLLKCLHNNLCQKYSAHAFQFSPRNVLWLLVVCLRSDNKRPCLEFSQLSVKDSFRDLFIPRIETILMMYTRNNLNCAEPLFEQNNSLNVNFNTQKKTVWLIHGYRPVGSIPLWLQNFVRILLNEEDMNVIVVDWSRGATTFIYNRAVKNTRKVAVSLSVHIKNLLKHGASLDNFHFIGVSLGAHISGFVGKIFHGQLGRITGLDPAGPRFSRKPPYSRLDYTDAKFVDVIHSDSNGLGIQEPLGHIDFYPNGGNKQPGCPKSIFSGIQFIKCNHQRAVHLFMASLETNCNFISFPCRSYKDYKTSLCVDCDCFKEKSCPRLGYQAKLFKGVLKERMEGRPLRTTVFLDTSGTYPFCTYYFVLSIIVPDKTMMDGSFSFKLLNQLGMIEEPRLYEKNKPFYKLQEVKILAQFYNDFVNISSIGLTYFQSSNLQCSTCTYKIQRLMLKSLTYPERPPLCRYNIVLKDREEVFLNPNTCTPKNTEspecificaciones
AAI48692.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.86kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LPDL/PRED5 | |
LIPI | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
149998 | |
LIPI (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLLKCLHNNLCQKYSAHAFQFSPRNVLWLLVVCLRSDNKRPCLEFSQLSVKDSFRDLFIPRIETILMMYTRNNLNCAEPLFEQNNSLNVNFNTQKKTVWLIHGYRPVGSIPLWLQNFVRILLNEEDMNVIVVDWSRGATTFIYNRAVKNTRKVAVSLSVHIKNLLKHGASLDNFHFIGVSLGAHISGFVGKIFHGQLGRITGLDPAGPRFSRKPPYSRLDYTDAKFVDVIHSDSNGLGIQEPLGHIDFYPNGGNKQPGCPKSIFSGIQFIKCNHQRAVHLFMASLETNCNFISFPCRSYKDYKTSLCVDCDCFKEKSCPRLGYQAKLFKGVLKERMEGRPLRTTVFLDTSGTYPFCTYYFVLSIIVPDKTMMDGSFSFKLLNQLGMIEEPRLYEKNKPFYKLQEVKILAQFYNDFVNISSIGLTYFQSSNLQCSTCTYKIQRLMLKSLTYPERPPLCRYNIVLKDREEVFLNPNTCTPKNT | |
RUO | |
LIPI | |
Wheat Germ (in vitro) | |
GST | |
Liquid |