Learn More
Abnova™ Human LIG3 Partial ORF (NP_039269, 823 a.a. - 922 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003980-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the DNA ligase family. Each member of this family encodes a protein that catalyzes the joining of DNA ends but they each have a distinct role in DNA metabolism. The protein encoded by this gene is involved in excision repair and is located in both the mitochondria and nucleus, with translation initiation from the upstream start codon allowing for transport to the mitochondria and translation initiation from a downstream start codon allowing for transport to the nucleus. Additionally, alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: TKSSPVKVGEKRKAADETLCQTKVLLDIFTGVRLYLPPSTPDFSRLRRYFVAFDGDLVQEFDMTSATHVLGSRDKNPAAQQVSPEWIWACIRKRRLVAPCEspecificaciones
NP_039269 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TKSSPVKVGEKRKAADETLCQTKVLLDIFTGVRLYLPPSTPDFSRLRRYFVAFDGDLVQEFDMTSATHVLGSRDKNPAAQQVSPEWIWACIRKRRLVAPC | |
RUO | |
LIG3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3980 | |
LIG3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LIG2 | |
LIG3 | |
Recombinant | |
wheat germ expression system |