missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LALBA Partial ORF (NP_002280, 33 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003906-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16170565

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes alpha-lactalbumin, a principal protein of milk. Alpha-lactalbumin forms the regulatory subunit of the lactose synthase (LS) heterodimer and beta 1,4-galactosyltransferase (beta4Gal-T1) forms the catalytic component. Together, these proteins enable LS to produce lactose by transferring galactose moieties to glucose. As a monomer, alpha-lactalbumin strongly binds calcium and zinc ions and may possess bactericidal or antitumor activity. A folding variant of alpha-lactalbumin, called HAMLET, likely induces apoptosis in tumor and immature cells. [provided by RefSeq]

Sequence: DIDGYGGIALPELICTMFHTSGYDTQAIVENNESTEYGLFQISNKLWCKSSQVPQSRNICDISCDKFLDDDITDDIMCAKKILDIKGIDYWLAHKALCTEKLEQWLCEKL
Especificaciones

Especificaciones

NP_002280
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DIDGYGGIALPELICTMFHTSGYDTQAIVENNESTEYGLFQISNKLWCKSSQVPQSRNICDISCDKFLDDDITDDIMCAKKILDIKGIDYWLAHKALCTEKLEQWLCEKL
RUO
LALBA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3906
LALBA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138521/MGC138523
LALBA
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales