Learn More
Abnova™ Human LAIR1 Full-length ORF (AAH27899, 1 a.a. - 287 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003903-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is an inhibitory receptor found on peripheral mononuclear cells, including NK cells, T cells, and B cells. Inhibitory receptors regulate the immune response to prevent lysis of cells recognized as self. The gene is a member of both the immunoglobulin superfamily and the leukocyte-associated inhibitory receptor family. The gene maps to a region of 19q13.4 called the leukocyte receptor cluster, which contains at least 29 genes encoding leukocyte-expressed receptors of the immunoglobulin superfamily. [provided by RefSeq]
Sequence: MSPHPTALLGLVLCLAQTIHTQEEDLPRPSISAEPGTVIPLGSHVTFVCRGPVGVQTFRLERESRSTYNDTEDVSQASPSESEARFRIDSVSEGNAGPYRCIYYKPPKWSEQSDYLELLVKETSGGPDSPDTEPGSSAGPTQRPSDNSHNEHAPASQGLKAEHLYILIGVSVVFLFCLLLLVLFCLHRQNQIKQGPPRSKDEEQKPQQRPDLAVDVLERTADKATVNGLPEKDRETDTSALAAGSSQEVTYAQLDHWALTQRTARAVSPQSTKPMAESITYAAVARHEspecificaciones
AAH27899 | |
Liquid | |
3903 | |
LAIR1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSPHPTALLGLVLCLAQTIHTQEEDLPRPSISAEPGTVIPLGSHVTFVCRGPVGVQTFRLERESRSTYNDTEDVSQASPSESEARFRIDSVSEGNAGPYRCIYYKPPKWSEQSDYLELLVKETSGGPDSPDTEPGSSAGPTQRPSDNSHNEHAPASQGLKAEHLYILIGVSVVFLFCLLLLVLFCLHRQNQIKQGPPRSKDEEQKPQQRPDLAVDVLERTADKATVNGLPEKDRETDTSALAAGSSQEVTYAQLDHWALTQRTARAVSPQSTKPMAESITYAAVARH | |
RUO | |
LAIR1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.31kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD305/LAIR-1 | |
LAIR1 | |
Yes | |
wheat germ expression system |