Learn More
Abnova™ Human LACE1 Full-length ORF (NP_660358.2, 1 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00246269-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with possible ATPase function. The protein contains a P-loop motif and a five-domain structure that is conserved in fly, yeast, and bacteria. Two conserved estrogen receptor binding sites are located within 2.5 kb of this gene. [provided by RefSeq]
Sequence: MAASWSLLVTLRPLAQSPLRGRCVGCGAWAAALAPLATAPGKPFWKAYTVQTSESMTPTATSETYLKALAVCHGPLDHYDFLIKAHELKDDEHQRRVIQCLQKLHEDLKGYNIEAEGLFSKLFSRSKPPRGLYVYGDVGTGKTMVMDMFYAYVEMKRKKRVHFHGFMLDVHKRIHRLKQSLPKRKPGFMAKSYDPIAPIAEEISEEACLLCFDEFQVTDIADAMILKQLFENLFKNGVVVVATSNRPPEDLYKNGLQRANFVPFIAVLKEYCNTVQLDSGIDYRKRELPAAGKLYYLTSEADVEAVMDKLFDELAQKQNDLTRPRILKVQGRELRLNKACGTVADCTFEELCERPLGASDYLELSKNFDTIFLRNIPQFTLANRTQGRRFITLIDNFYDLKVRIICSASTPISSLFLHQHHDSELEQSRILMDDLGLSQDSAEGLSMFTGEEEIFAFQRTISRLTEMQTEQYWNEGDRTKKEspecificaciones
NP_660358.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AFG1 | |
LACE1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
246269 | |
LACE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAASWSLLVTLRPLAQSPLRGRCVGCGAWAAALAPLATAPGKPFWKAYTVQTSESMTPTATSETYLKALAVCHGPLDHYDFLIKAHELKDDEHQRRVIQCLQKLHEDLKGYNIEAEGLFSKLFSRSKPPRGLYVYGDVGTGKTMVMDMFYAYVEMKRKKRVHFHGFMLDVHKRIHRLKQSLPKRKPGFMAKSYDPIAPIAEEISEEACLLCFDEFQVTDIADAMILKQLFENLFKNGVVVVATSNRPPEDLYKNGLQRANFVPFIAVLKEYCNTVQLDSGIDYRKRELPAAGKLYYLTSEADVEAVMDKLFDELAQKQNDLTRPRILKVQGRELRLNKACGTVADCTFEELCERPLGASDYLELSKNFDTIFLRNIPQFTLANRTQGRRFITLIDNFYDLKVRIICSASTPISSLFLHQHHDSELEQSRILMDDLGLSQDSAEGLSMFTGEEEIFAFQRTISRLTEMQTEQYWNEGDRTKK | |
RUO | |
LACE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |