missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KSR Partial ORF (XP_290793, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008844-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: LDARRESGSGPSTDTLSAASLPWPPGSSQLGRAGNSAQGPRSISVSALPASDSPTPSFSEGLSDTCIPLHASGRLTPRALHSFITPPTTPQLRRHTKLKPEspecificaciones
XP_290793 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LDARRESGSGPSTDTLSAASLPWPPGSSQLGRAGNSAQGPRSISVSALPASDSPTPSFSEGLSDTCIPLHASGRLTPRALHSFITPPTTPQLRRHTKLKP | |
RUO | |
KSR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8844 | |
KSR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KSR/RSU2 | |
KSR1 | |
Recombinant | |
wheat germ expression system |