Learn More
Abnova™ Human KRT34 Full-length ORF (AAH41070.1, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003885-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]
Sequence: MLYAKPPPTINGIKGLQRKERLKPAHIHLQQLTCFSITCSSTMSYSCCLPSLGCRTSCSSRPCVPPSCHGYTLPGACNIPANVSNCNWFCEGSFNGSEKETMQFLNDRLASYLEKVRQLERDNAELEKLIQERSQQQEPLLCPSYQSYFKTIEELQQKILCAKAENARLVVNIDNAKLASDDFRSKYQTEQSLRLLVESDINSIRRILDELTLCKSDLESQVESLREELICLKKNHEEEVNTLRSQLGDRLNVEVDTAPTVDLNQVLNETRSQYEALVETNRREVEQWFATQTEELNKQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHNLRDSLENTLTESEAHYSSQLSQVQSLITNVESQLAEIRCDLERQNQEYQVLLDVRARLECEINTYRSLLESEDCKLPCNPCATTNASGNSCGPCGTSQKGCCNEspecificaciones
AAH41070.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HA4/Ha-4/KRTHA4/hHa4 | |
KRT34 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3885 | |
KRT34 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLYAKPPPTINGIKGLQRKERLKPAHIHLQQLTCFSITCSSTMSYSCCLPSLGCRTSCSSRPCVPPSCHGYTLPGACNIPANVSNCNWFCEGSFNGSEKETMQFLNDRLASYLEKVRQLERDNAELEKLIQERSQQQEPLLCPSYQSYFKTIEELQQKILCAKAENARLVVNIDNAKLASDDFRSKYQTEQSLRLLVESDINSIRRILDELTLCKSDLESQVESLREELICLKKNHEEEVNTLRSQLGDRLNVEVDTAPTVDLNQVLNETRSQYEALVETNRREVEQWFATQTEELNKQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHNLRDSLENTLTESEAHYSSQLSQVQSLITNVESQLAEIRCDLERQNQEYQVLLDVRARLECEINTYRSLLESEDCKLPCNPCATTNASGNSCGPCGTSQKGCCN | |
RUO | |
KRT34 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |