missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KIAA1333 Partial ORF (NP_060239, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055632-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: NESKPGDSQNLACVFCRKHDDCPNKYGEKKTKEKWNLTVHYYCLLMSSGIWQRGKEEEGVYGFLIEDIRKEVNRASKLKCCVCKKNGASIGCVAPRCKREspecificaciones
NP_060239 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NESKPGDSQNLACVFCRKHDDCPNKYGEKKTKEKWNLTVHYYCLLMSSGIWQRGKEEEGVYGFLIEDIRKEVNRASKLKCCVCKKNGASIGCVAPRCKR | |
RUO | |
KIAA1333 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55632 | |
KIAA1333 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20333/G2E3/PHF7B | |
KIAA1333 | |
Recombinant | |
wheat germ expression system |