Learn More
Abnova™ Human KCNK1 Partial ORF (NP_002236.1, 265 a.a. - 336 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003775-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq]
Sequence: ETFCELHELKKFRKMFYVKKDKDEDQVHIIEHDQLSFSSITDQAAGMKEDQKQNEPFVATQSSACVDGPANHEspecificaciones
NP_002236.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETFCELHELKKFRKMFYVKKDKDEDQVHIIEHDQLSFSSITDQAAGMKEDQKQNEPFVATQSSACVDGPANH | |
RUO | |
KCNK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3775 | |
KCNK1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DPK/HOHO/K2p1.1/KCNO1/TWIK-1/TWIK1 | |
KCNK1 | |
Recombinant | |
wheat germ expression system |