missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JRK Partial ORF (NP_003715, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008629-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16118915

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 25-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is the human homolog of the mouse jerky gene. The encoded protein has similarity to several nuclear regulatory proteins, including centromere protein B, suggesting that it might function as a DNA-binding protein. Insertional inactivation of this gene in transgenic mice resulted in epileptic seizures. Childhood Absence Epilepsy (CAE) has been mapped to the same chromosomal location (8q24.3) as this gene, making this gene a strong candidate for CAE. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGK
Especificaciones

Especificaciones

NP_003715
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGK
RUO
JRK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8629
JRK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686C24207/FLJ45729/JH8
JRK
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales