missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human JMJD1C Partial ORF (NP_004232, 2 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00221037-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: IVMNDQVLEPQNVDPSMVQMTFLDDVVHSLLKGENIGITSRRRSRANQNVNAVHSHYTRAQANSPRPAMNSQAAVPKQNTHQQQQQRSIRPNKRKGSDEspecificaciones
NP_004232 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IVMNDQVLEPQNVDPSMVQMTFLDDVVHSLLKGENIGITSRRRSRANQNVNAVHSHYTRAQANSPRPAMNSQAAVPKQNTHQQQQQRSIRPNKRKGSD | |
RUO | |
JMJD1C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
221037 | |
JMJD1C (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761F0118/FLJ14374/KIAA1380/RP11-10C13.2/TRIP8 | |
JMJD1C | |
Recombinant | |
wheat germ expression system |