Learn More
Abnova™ Human ITSN2 Partial ORF (NP_006268, 471 a.a. - 571 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00050618-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a cytoplasmic protein which contains SH3 domains. This protein is a member of a family of proteins involved in clathrin-mediated endocytosis. Intersectin 2 is thought to regulate the formation of clathrin-coated vesicles and also may function in the induction of T cell antigen receptor (TCR) endocytosis. Alternatively spliced transcript variants have been found for this gene that encode three distinct isoforms. Additional variants have been found but their full length nature has not been determined. [provided by RefSeq]
Sequence: NQKNREQEEIVRLNSKKKNLHLELEALNGKHQQISGRLQDVRLKKQTQKTELEVLDKQCDLEIMEIKQLQQELQEYQNKLIYLVPEKQLLNERIKNMQFSNEspecificaciones
NP_006268 | |
Liquid | |
50618 | |
ITSN2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1256/PRO2015/SH3D1B/SH3P18/SWA/SWAP | |
ITSN2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NQKNREQEEIVRLNSKKKNLHLELEALNGKHQQISGRLQDVRLKKQTQKTELEVLDKQCDLEIMEIKQLQQELQEYQNKLIYLVPEKQLLNERIKNMQFSN | |
RUO | |
ITSN2 | |
Wheat Germ (in vitro) | |
GST |