Learn More
ACROBiosystems Human IL-27 Ra / TCCR Protein, Fc Tag
Marca: ACROBiosystems ILA-H5254-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Interleukin-27 receptor subunit alpha ( IL27RA) is also known as Cytokine receptor WSX-1, Cytokine receptor-like 1, Type I T-cell cytokine receptor (TCCR), ZcytoR1, IL27RA belongs to the type I cytokine receptor family and type 2 subfamily. IL27RA contains three fibronectin type-III domains. IL27RA is highly expressed in lymphoid tissues such as spleen, lymph nodes and peripheral blood leukocytes. IL27RA is receptor for IL27, but requires IL6ST/gp130 to mediate signal transduction in response to IL27. IL27RA involved in the regulation of Th1-type immune responses. IL27RA also appears to be involved in innate defense mechanisms.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 78.9 kDa. The protein migrates as 100-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH28003 | |
78.9 kDa | |
1 mg | |
Gln 33 - Arg 514 | |
IL27RA,CRL1,TCCR,WSX1 | |
Unconjugated | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human IL-27 Ra, Fc Tag (ILA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Arg 514 (Accession # AAH28003). | |
-20°C | |
IL-27 Ra | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |