Learn More
ACROBiosystems Human IL-23 R Protein, Fc Tag
Marca: ACROBiosystems ILR-H5254-100ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Interleukin 23 receptor (IL-23R) is a type I cytokine receptor, and IL-23R pairs with the receptor molecule IL12RB1/IL12Rbeta1, and both are required for IL23A signaling. Also, IL-23R associates constitutively with Janus kinase 2 (JAK2), and binds to transcription activator STAT3 in a ligand-dependent manner. Furthermore, IL-23R mediates T-cells, NK cells and possibly certain macrophage/myeloid cells stimulation probably through activation of the Jak-Stat signaling cascade. As for IL-23, it may be responsible for autoimmune inflammatory diseases and be important for tumorigenesis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_653302.2 | |
100 μg | |
Gly 24 - Gly 355 | |
IL-23 R,IL-23 Receptor | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
64.5 kDa | |
Human IL-23 R, Fc Tag (ILR-H5254) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 355 (Accession # Q5VWK5-1). | |
-20°C | |
IL-23 R | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |