Learn More
ACROBiosystems Human IL-2 R beta&IL-2 R gamma Heterodimer Protein, Fc Tag&Fc Tag
MALS verified
Marca:  ACROBiosystems ILG-H5254-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17196441
  • 3108.82€ / 1 miligramo
Fecha estimada de envío: 08-05-2024
para ver el stock
Descripción
Descripción
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.
Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of IL-2RB and IL-2RG, has a calculated MW of 51.1 kDa (IL-2RB) and 53.6 kDa (IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-2RG
Especificaciones
Especificaciones
NP_000869.1 (IL-2 RB) and AAH14972.1 (IL-2 RG)
1 mg
Ala 27 - Asp 239 (IL-2 RB) and Leu 23 - Asn 254 (IL-2 RG)
IL-2 R beta and IL-2 R gamma,IL-2 RB and IL-2 RG
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
51.1 kDa (IL-2 RB) and 53.6 kDa (IL-2 RG)
Human IL-2 RB&IL-2; RG Heterodimer Protein, Fc Tag&Fc; Tag (MALS verified) (ILG-H5254) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (IL-2 RB) and Leu 23 - Asn 254 (IL-2 RG) (Accession # P14784-1 (IL-2 RB) and P31785-1 (IL-2 RG)).
-20°C
IL-2 R beta and IL-2 R gamma
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens