Learn More
ACROBiosystems Human IL-2 R beta / CD122 Protein, His Tag
SPR verified
Marca: ACROBiosystems CD2-H5221-100ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Interleukin-2 receptor (IL-2R) is a heterotrimeric protein expressed on the surface of certain immune cells, such as lymphocytes, that binds and responds to a cytokine called IL-2. The IL-2R is made up of 3 subunits - α (CD25), β (CD122) and γ (CD132) - non-covalently associating. The α and β chains are involved in binding IL-2, while signal transduction following cytokine interaction is carried out by the γ-chain, along with the β subunit. CD122 is also known as IL2R beta, is a member of the type I cytokine receptor family. CD122 is the receptor for interleukin-2 and is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_000869.1 | |
100 μg | |
Ala 27 - Asp 239 | |
IL2RB,RP5-1170K4.6,CD122,P70-75 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
25.4 kDa | |
Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # NP_000869.1). | |
-20°C | |
IL-2 R beta | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |