Learn More
Abnova™ Human IGSF1 Partial ORF (NP_001546, 220 a.a. - 310 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003547-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the immunoglobulin (Ig) superfamily (see MIM 147100), which includes IGSF1, have a variety of functions, but all appear to play a role in cell recognition and the regulation of cell behavior.[supplied by OMIM]
Sequence: VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSDEspecificaciones
NP_001546 | |
Liquid | |
3547 | |
IGSF1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGCD1/IGDC1/INHBP/KIAA0364/MGC75490/PGSF2 | |
IGSF1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSD | |
RUO | |
IGSF1 | |
Wheat Germ (in vitro) | |
GST |