missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HTR1E Partial ORF (NP_000856.1, 206 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003354-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: YHAAKSLYQKRGSSRHLSNRSTDSQNSFASCKLTQTFCVSDFSTSDPTTEFEKFHASIRIPPFDNDLDHPGEspecificaciones
NP_000856.1 | |
Liquid | |
3354 | |
HTR1E (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5-HT1E | |
HTR1E | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YHAAKSLYQKRGSSRHLSNRSTDSQNSFASCKLTQTFCVSDFSTSDPTTEFEKFHASIRIPPFDNDLDHPG | |
RUO | |
HTR1E | |
Wheat Germ (in vitro) | |
GST |