missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSPA1L Full-length ORF (AAH34483.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003305-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166431

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a 70kDa heat shock protein. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which also encode isoforms of the 70kDa heat shock protein. [provided by RefSeq]

Sequence: MATAKGIAIGIDLGTTYSCVGVFQHGKVEIIANDQGNRTTPSYVAFTDTERLIGDAAKNQVAMNPQNTVFDAKRLIGRKFNDPVVQADMKLWPFQVINEGGKPKVLVSYKGENKAFYPEEISSMVLTKLKETAEAFLGHPVTNAVITVPAYFNDSQRQATKDAGVIAGLNVLRIINEPTAAAIAYGLDKGGQGERHVLIFDLGGGTFDVSILTIDDGIFEVKATAGDTHLGGEDFDNRLVSHFVEEFKRKHKKDISQNKRAVRRLRTACERAKRTLSSSTQANLEIDSLYEGIDFYTSITRARFEELCADLFRGTLEPVEKALRDAKMDKAKIHDIVLVGGSTRIPKVQRLLQDYFNGRDLNKSINPDEAVAYGAAVQAAILMGDKSEKVQDLLLLDVAPLSLGLETAGGVMTALMKRNSTIPTKQTQIFTTYSDNQPGVLIQVYEGERAMTKDNNLLGRFDLTGIPPAPRGVPQIEVTFDIDANGILNVTAMDKSTGKVNKITIANDKGRLSKEEIERMVLDAEKYKAEDEVQREKIAAKNALESYAFNMKSVVSDEGLKGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD
Especificaciones

Especificaciones

AAH34483.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSP70-1L/HSP70-HOM/HSP70T/hum70t
HSPA1L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3305
HSPA1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATAKGIAIGIDLGTTYSCVGVFQHGKVEIIANDQGNRTTPSYVAFTDTERLIGDAAKNQVAMNPQNTVFDAKRLIGRKFNDPVVQADMKLWPFQVINEGGKPKVLVSYKGENKAFYPEEISSMVLTKLKETAEAFLGHPVTNAVITVPAYFNDSQRQATKDAGVIAGLNVLRIINEPTAAAIAYGLDKGGQGERHVLIFDLGGGTFDVSILTIDDGIFEVKATAGDTHLGGEDFDNRLVSHFVEEFKRKHKKDISQNKRAVRRLRTACERAKRTLSSSTQANLEIDSLYEGIDFYTSITRARFEELCADLFRGTLEPVEKALRDAKMDKAKIHDIVLVGGSTRIPKVQRLLQDYFNGRDLNKSINPDEAVAYGAAVQAAILMGDKSEKVQDLLLLDVAPLSLGLETAGGVMTALMKRNSTIPTKQTQIFTTYSDNQPGVLIQVYEGERAMTKDNNLLGRFDLTGIPPAPRGVPQIEVTFDIDANGILNVTAMDKSTGKVNKITIANDKGRLSKEEIERMVLDAEKYKAEDEVQREKIAAKNALESYAFNMKSVVSDEGLKGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD
RUO
HSPA1L
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales