Learn More
Abnova™ Human HSN2 Full-length ORF (NP_998820.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00378465-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This intronless gene is poorly expressed and lies within an intron of WNK1 gene (GeneID:65125) on chr 12. Mutations in this gene are associated with hereditary sensory and autonomic neuropathy, type II, an autosomal recessive disorder characterized by impairment of pain, temperature, and touch sensation owing to reduction or absence of peripheral sensory neurons. It is proposed that this gene product may play a role in the development and/or maintenance of peripheral sensory neurons or their supporting Schwann cells. [provided by RefSeq]
Sequence: MYELLVLFMLIQPQSMAHPCGGTPTYPESQIFFPTIHERPVSFSPPPTCPPKVAISQRRKSTSFLEAQTHHFQPLLRTVGQSLLPPGGSPTNWTPEAVVMLGTTASRVTGESCEIQVHPMFEPSQVYSDYRPGLVLPEEAHYFIPQEAVYVAGVHYQARVAEQYEGIPYNSSVLSSPMKQIPEQKPVQGGPTSSSVFEFPSGQAFLVGHLQNLRLDSGLGPGSPLSSISAPISTDATRLKFHPVFVPHSAPAVLTHNNESRSNCVFEFHVHTPSSSSGEGGGILPQRVYRNRQVAVDLNQEELPPQSVGLHGYLQPVTEEKHNYHAPELTVSVVEPIGQNWPIGSPEYSSDSSQITSSDPSDFQSPPPTGGAAAPFGSDVSMPFIHLPQTVLQESPLFFCFPQGTTSQQVLTASFSSGGSALHPQVIGKLPQLFEspecificaciones
NP_998820.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSAN2 | |
HSN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
378465 | |
HSN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYELLVLFMLIQPQSMAHPCGGTPTYPESQIFFPTIHERPVSFSPPPTCPPKVAISQRRKSTSFLEAQTHHFQPLLRTVGQSLLPPGGSPTNWTPEAVVMLGTTASRVTGESCEIQVHPMFEPSQVYSDYRPGLVLPEEAHYFIPQEAVYVAGVHYQARVAEQYEGIPYNSSVLSSPMKQIPEQKPVQGGPTSSSVFEFPSGQAFLVGHLQNLRLDSGLGPGSPLSSISAPISTDATRLKFHPVFVPHSAPAVLTHNNESRSNCVFEFHVHTPSSSSGEGGGILPQRVYRNRQVAVDLNQEELPPQSVGLHGYLQPVTEEKHNYHAPELTVSVVEPIGQNWPIGSPEYSSDSSQITSSDPSDFQSPPPTGGAAAPFGSDVSMPFIHLPQTVLQESPLFFCFPQGTTSQQVLTASFSSGGSALHPQVIGKLPQLF | |
RUO | |
HSN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |