Learn More
Abnova™ Human HSD17B12 Partial ORF (NP_057226, 203 a.a. - 271 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051144-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.[supplied by OMIM]
Sequence: SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNGEspecificaciones
NP_057226 | |
Liquid | |
51144 | |
HSD17B12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KAR/SDR12C1 | |
HSD17B12 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNG | |
RUO | |
HSD17B12 | |
Wheat Germ (in vitro) | |
GST |