missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HRMT1L2 Partial ORF (NP_001527, 260 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003276-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The PRMT1 gene encodes a protein arginine methyltransferase that functions as a histone methyltransferase specific for H4 (see MIM 602822).[supplied by OMIM]
Sequence: PFCLQVKRNDYVHALVAYFNIEFTRCHKRTGFSTSPESPYTHWKQTVFYMEDYLTVKTGEEIFGTIGMRPNAKNNRDLDFTIDLDFKGQLCELSCSTDYRMREspecificaciones
NP_001527 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PFCLQVKRNDYVHALVAYFNIEFTRCHKRTGFSTSPESPYTHWKQTVFYMEDYLTVKTGEEIFGTIGMRPNAKNNRDLDFTIDLDFKGQLCELSCSTDYRMR | |
RUO | |
PRMT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3276 | |
HRMT1L2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANM1/HCP1/HRMT1L2/IR1B4 | |
PRMT1 | |
Recombinant | |
wheat germ expression system |