missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HLCS Full-length ORF (NP_000402.3, 1 a.a. - 726 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003141-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196191

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 06-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Holocarboxylase synthetase (EC 6.3.4.10) covalently links biotin to propionyl-CoA carboxylase (PCCA; MIM 232000), pyruvate carboxylase (PC; MIM 608786), alpha-methylcrotonyl-CoA carboxylase (MCCC1; MIM 609010), and acetyl-CoA carboxylase (ACACA; MIM 200350).[supplied by OMIM]

Sequence: MEDRLHMDNGLVPQKIVSVHLQDSTLKEVKDQVSNKQAQILEPKPEPSLEIKPEQDGMEHVGRDDPKALGEEPKQRRGSASGSEPAGDSDRGGGPVEHYHLHLSSCHECLELENSTIESVKFASAENIPDLPYDYSSSLESVADETSPEREGRRVNLTGKAPNILLYVGSDSQEALGRFHEVRSVLADCVDIDSYILYHLLEDSALRDPWTDNCLLLVIATRESIPEDLYQKFMAYLSQGGKVLGLSSSFTFGGFQVTSKGALHKTVQNLVFSKADQSEVKLSVLSSGCRYQEGPVRLSPGRLQGHLENEDKDRMIVHVPFGTRGGEAVLCQVHLELPPSSNIVQTPEDFNLLKSSNFRRYEVLREILTTLGLSCDMKQVPALTPLYLLSAAEEIRDPLMQWLGKHVDSEGEIKSGQLSLRFVSSYVSEVEITPSCIPVVTNMEAFSSEHFNLEIYRQNLQTKQLGKVILFAEVTPTTMRLLDGLMFQTPQEMGLIVIAARQTEGKGRGGNVWLSPVGCALSTLLISIPLRSQLGQRIPFVQHLMSVAVVEAVRSIPEYQDINLRVKWPNDIYYSDLMKIGGVLVNSTLMGETFYILIGCGFNVTNSNPTICINDLITEYNKQHKAELKPLRADYLIARVVTVLEKLIKEFQDKGPNSVLPLYYRYWVHSGQQVHLGSAEGPKVSIVGLDDSGFLQVHQEGGEVVTVHPDGNSFDMLRNLILPKRR
Especificaciones

Especificaciones

NP_000402.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
107.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HCS
HLCS
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3141
HLCS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEDRLHMDNGLVPQKIVSVHLQDSTLKEVKDQVSNKQAQILEPKPEPSLEIKPEQDGMEHVGRDDPKALGEEPKQRRGSASGSEPAGDSDRGGGPVEHYHLHLSSCHECLELENSTIESVKFASAENIPDLPYDYSSSLESVADETSPEREGRRVNLTGKAPNILLYVGSDSQEALGRFHEVRSVLADCVDIDSYILYHLLEDSALRDPWTDNCLLLVIATRESIPEDLYQKFMAYLSQGGKVLGLSSSFTFGGFQVTSKGALHKTVQNLVFSKADQSEVKLSVLSSGCRYQEGPVRLSPGRLQGHLENEDKDRMIVHVPFGTRGGEAVLCQVHLELPPSSNIVQTPEDFNLLKSSNFRRYEVLREILTTLGLSCDMKQVPALTPLYLLSAAEEIRDPLMQWLGKHVDSEGEIKSGQLSLRFVSSYVSEVEITPSCIPVVTNMEAFSSEHFNLEIYRQNLQTKQLGKVILFAEVTPTTMRLLDGLMFQTPQEMGLIVIAARQTEGKGRGGNVWLSPVGCALSTLLISIPLRSQLGQRIPFVQHLMSVAVVEAVRSIPEYQDINLRVKWPNDIYYSDLMKIGGVLVNSTLMGETFYILIGCGFNVTNSNPTICINDLITEYNKQHKAELKPLRADYLIARVVTVLEKLIKEFQDKGPNSVLPLYYRYWVHSGQQVHLGSAEGPKVSIVGLDDSGFLQVHQEGGEVVTVHPDGNSFDMLRNLILPKRR
RUO
HLCS
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales