missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HIC1 Partial ORF (NP_006488.2, 627 a.a. - 705 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003090-Q02.10ug
Detalles adicionales : Peso : 0.02000kg
Descripción
Sequence: PEGVFAVARLTAEQLSLKQQDKAAAAELLAQTTHFLHDPKVALESLYPLAKFTAELGLSPDKAAEVLSQGAHLAAGPDGEspecificaciones
NP_006488.2 | |
Liquid | |
3090 | |
HIC1 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZBTB29/hic-1 | |
HIC1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.32kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PEGVFAVARLTAEQLSLKQQDKAAAAELLAQTTHFLHDPKVALESLYPLAKFTAELGLSPDKAAEVLSQGAHLAAGPDG | |
RUO | |
HIC1 | |
Wheat Germ (in vitro) | |
GST |