Learn More
Abnova™ Human HEBP1 Full-length ORF (AAH16277.1, 1 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00050865-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells. [provided by RefSeq]
Sequence: MLGMIKNSLFGSVETWPWQVLSKGDKEEVAYEERACEGGKFATVEVTDKPVDEALREAMPKVAKYAGGTNDKGIGMGMTVPISFAVFPNEDGSLQKKLKVWFRIPNQFQSDPPAPSDKSVKIEEREGITVYSMQFGGYAKEADYVAQATRLRAALEGTATYRGDIYFCTGYDPPMKPYGRRNEIWLLKTEspecificaciones
AAH16277.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBP/HEBP | |
HEBP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
50865 | |
HEBP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGMIKNSLFGSVETWPWQVLSKGDKEEVAYEERACEGGKFATVEVTDKPVDEALREAMPKVAKYAGGTNDKGIGMGMTVPISFAVFPNEDGSLQKKLKVWFRIPNQFQSDPPAPSDKSVKIEEREGITVYSMQFGGYAKEADYVAQATRLRAALEGTATYRGDIYFCTGYDPPMKPYGRRNEIWLLKT | |
RUO | |
HEBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |