missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HAPLN2 Partial ORF (NP_068589, 31 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00060484-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: HPGPHYLLPPIHEVIHSHRGATATLPCVLGTTPPSYKVRWSKVEPGELRETLILITNGLHARGYGPLGGRARMRRGHRLDASLVIAGVRLEDEGRYRCELINGIEDESVALTEspecificaciones
NP_068589 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.06kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HPGPHYLLPPIHEVIHSHRGATATLPCVLGTTPPSYKVRWSKVEPGELRETLILITNGLHARGYGPLGGRARMRRGHRLDASLVIAGVRLEDEGRYRCELINGIEDESVALT | |
RUO | |
HAPLN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
60484 | |
HAPLN2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRAL1 | |
HAPLN2 | |
Recombinant | |
wheat germ expression system |