Learn More
Abnova™ Human GZMA Full-length ORF (NP_006135.1, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003001-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface 'nonself' antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells. [provided by RefSeq]
Sequence: MRNSYRFLASSLSVVVSLLLIPEDVCEKIIGGNEVTPHSRPYMVLLSLDRKTICAGALIAKDWVLTAAHCNLNKRSQVILGAHSITREEPTKQIMLVKKEFPYPCYDPATREGDLKLLQLTEKAKINKYVTILHLPKKGDDVKPGTMCQVAGWGRTHNSASWSDTLREVNITIIDRKVCNDRNHYNFNPVIGMNMVCAGSLRGGRDSCNGDSGSPLLCEGVFRGVTSFGLENKCGDPRGPGVYILLSKKHLNWIIMTIKGAVEspecificaciones
NP_006135.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTLA3/HFSP | |
GZMA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3001 | |
GZMA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRNSYRFLASSLSVVVSLLLIPEDVCEKIIGGNEVTPHSRPYMVLLSLDRKTICAGALIAKDWVLTAAHCNLNKRSQVILGAHSITREEPTKQIMLVKKEFPYPCYDPATREGDLKLLQLTEKAKINKYVTILHLPKKGDDVKPGTMCQVAGWGRTHNSASWSDTLREVNITIIDRKVCNDRNHYNFNPVIGMNMVCAGSLRGGRDSCNGDSGSPLLCEGVFRGVTSFGLENKCGDPRGPGVYILLSKKHLNWIIMTIKGAV | |
RUO | |
GZMA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |