missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GTF2IRD2 Full-length ORF (AAH67859.1, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084163-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16165673

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is unusual in that its coding sequence is mostly derived from Charlie8 repeat elements. However, there is mRNA and EST evidence to suggest that this gene is transcribed, and the encoded protein has a homolog in mouse, with which it shares 78% sequence identity. The exact function of this gene product is not known. It is inferred to be a transcription factor based on the presence of GTF2I-like repeats (containing helix-loop-helix motifs), also found in other proteins such as GTF2IRD1 and GTF2I. These three genes are clustered together on chr 7q11.23. [provided by RefSeq]

Sequence: MAQVAVSTLPVEEESSSETRMVVTFLVSALESMCKELAKSKAEVACIAVYETDVFVVGTERGCAFVNARTDFQKDFAKYCVAEGLCEVKPPCPVNGMQVHSGETEILRKAVEDYFCFCYGKALGTTVMVPVPYEKMLRDQSAVVVQGLPEGVAFQHPENYDLATLKWILENKAGISFIINRPFLGPESQLGGPGMVTDAERSIVSPSESCGPINVKTEPMEDSGISLKAEAVSVKKESEDPNYYQYNMQGSHPSSTSNEVIEMELPMEDSTPLVPSEEPNEDPEAEVKIEGNTNSSSVTNSAAGVEDLNIVQVTVPDNEKERLSSIEKIKQLREQVNDLFSRKFGEAIGVDFPVKVPYRKITFNPGCVVIDGMPPGVVFKAPGYLEISSMRRILEAAEFIKFTVIRPLPGLELSNGLLEAGAGGLLEAGSLRPAWATWQDPISTKNLKISWKQWLTPVAPATWQAEAGGSLELRSLRQQRTIIAILHSSLGDRTRPCL
Especificaciones

Especificaciones

AAH67859.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21423/FLJ37938/FP630/GTF2IRD2A/MGC75203
GTF2IRD2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84163
GTF2IRD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQVAVSTLPVEEESSSETRMVVTFLVSALESMCKELAKSKAEVACIAVYETDVFVVGTERGCAFVNARTDFQKDFAKYCVAEGLCEVKPPCPVNGMQVHSGETEILRKAVEDYFCFCYGKALGTTVMVPVPYEKMLRDQSAVVVQGLPEGVAFQHPENYDLATLKWILENKAGISFIINRPFLGPESQLGGPGMVTDAERSIVSPSESCGPINVKTEPMEDSGISLKAEAVSVKKESEDPNYYQYNMQGSHPSSTSNEVIEMELPMEDSTPLVPSEEPNEDPEAEVKIEGNTNSSSVTNSAAGVEDLNIVQVTVPDNEKERLSSIEKIKQLREQVNDLFSRKFGEAIGVDFPVKVPYRKITFNPGCVVIDGMPPGVVFKAPGYLEISSMRRILEAAEFIKFTVIRPLPGLELSNGLLEAGAGGLLEAGSLRPAWATWQDPISTKNLKISWKQWLTPVAPATWQAEAGGSLELRSLRQQRTIIAILHSSLGDRTRPCL
RUO
GTF2IRD2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales