Learn More
Abnova™ Human GRSF1 Partial ORF (NP_002083, 87 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002926-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a cellular protein that binds RNAs containing the G-rich element. The protein is localized in the cytoplasm, and has been shown to stimulate translation of viral mRNAs in vitro. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: EEEVDDVFLIRAQGLPWSCTMEDVLNFFSDCRIRNGENGIHFLLNRDGKRRGDALIEMESEQDVQKALEKHRMYMGQRYVEVYEINNEDVDALMKSLQEspecificaciones
NP_002083 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEEVDDVFLIRAQGLPWSCTMEDVLNFFSDCRIRNGENGIHFLLNRDGKRRGDALIEMESEQDVQKALEKHRMYMGQRYVEVYEINNEDVDALMKSLQ | |
RUO | |
GRSF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2926 | |
GRSF1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ13125 | |
GRSF1 | |
Recombinant | |
wheat germ expression system |