Learn More
ACROBiosystems Human Growth Hormone R (GHR) Protein, His Tag
Marca: ACROBiosystems GHR-H5222-50UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Growth hormone receptor (GHR) is also known as somatotropin receptor, growth hormone-binding protein (GHBR), which belongs to the type I cytokine receptor family or Type 1 subfamily. GHR contains one fibronectin type-III domain. GHR / GHBR is expressed in various tissues with high expression in liver and skeletal muscle. The soluble form (GHBP) is produced by phorbol ester-promoted proteolytic cleavage at the cell surface (shedding) by ADAM17/TACE. GHR is receptor for pituitary gland growth hormone involved in regulating postnatal body growth. On ligand binding, couples to the JAK2/STAT5 pathway. The soluble form (GHBP) acts as a reservoir of growth hormone in plasma and may be a modulator/inhibitor of GH signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_000154.1 | |
50 μg | |
Ala 27 - Tyr 264 | |
GHR,GHBP,GH receptor | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
29.6 kDa | |
Human GHR, His Tag (GHR-H5222) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912-1). | |
-20°C | |
Growth Hormone R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |