missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPR142 Full-length ORF (AAI52988.1, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00350383-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16170314

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 12-05-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

GPR142 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) (Fredriksson et al., 2003 [PubMed 14623098]).[supplied by OMIM]

Sequence: MSIMMLPMEQKIQWVPTSLQDITAVLGTEAYTEEDKSMVSHAQKSQHSCLSHSRWLRSPQVTGGSWDLRIRPSKDSSSFRQAQCLRKDPGANNHLESQGVRGTAGDADRELRGPSEKATAGQPRVTLLPTPHVSGLSQEFESHWPEIAERSPCVAGVIPVIYYSVLLGLGLPVSLLTAVALARLATRTRRPSYYYLLALTASDIIIQVVIVFAGFLLQGAVLARQVPQAVVRTANILEFAANHASVWIAILLTVDRYTALCHPLHHRAASSPGRTRRAIAAVLSAALLTGIPFYWWLDMWRDTDSPRTLDEVLKWAHCLTVYFIPCGVFLVTNSAIIHRLRRRGRSGLQPRVGKSTAILLGITTLFTLLWAPRVFVMLYHMYVAPVHRDWRVHLALDVANMVAMLHTAANFGLYCFVSKTFRATVRQVIHDAYLPCTLASQPEGMAAKPVMEPPGLPTGAEV
Especificaciones

Especificaciones

AAI52988.1
Liquid
350383
GPR142 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSIMMLPMEQKIQWVPTSLQDITAVLGTEAYTEEDKSMVSHAQKSQHSCLSHSRWLRSPQVTGGSWDLRIRPSKDSSSFRQAQCLRKDPGANNHLESQGVRGTAGDADRELRGPSEKATAGQPRVTLLPTPHVSGLSQEFESHWPEIAERSPCVAGVIPVIYYSVLLGLGLPVSLLTAVALARLATRTRRPSYYYLLALTASDIIIQVVIVFAGFLLQGAVLARQVPQAVVRTANILEFAANHASVWIAILLTVDRYTALCHPLHHRAASSPGRTRRAIAAVLSAALLTGIPFYWWLDMWRDTDSPRTLDEVLKWAHCLTVYFIPCGVFLVTNSAIIHRLRRRGRSGLQPRVGKSTAILLGITTLFTLLWAPRVFVMLYHMYVAPVHRDWRVHLALDVANMVAMLHTAANFGLYCFVSKTFRATVRQVIHDAYLPCTLASQPEGMAAKPVMEPPGLPTGAEV
RUO
GPR142
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.77kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PGR2
GPR142
Yes
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales