Learn More
Abnova™ Human GOLGA2 Partial ORF (AAH06381.1, 1 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002801-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined. [provided by RefSeq]
Sequence: EQAEARRQILETMQNDRTTISRALSQNRELKEQLAELQSGFVKLTNENMEITSALQSEQHVKRELGKKLGELQEKLSELKETVELKSQEAQSLQQQRDQYLGHLQQYVAAYQQLTSEKEVEspecificaciones
AAH06381.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.94kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EQAEARRQILETMQNDRTTISRALSQNRELKEQLAELQSGFVKLTNENMEITSALQSEQHVKRELGKKLGELQEKLSELKETVELKSQEAQSLQQQRDQYLGHLQQYVAAYQQLTSEKEV | |
RUO | |
GOLGA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2801 | |
GOLGA2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GM130/MGC20672 | |
GOLGA2 | |
Recombinant | |
wheat germ expression system |