Learn More
Abnova™ Human GNL1 Partial ORF (NP_005266, 111 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002794-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C. [provided by RefSeq]
Sequence: ELLELDIREVYQPGSVLDFPRRPPWSYEMSKEQLMSQEERSFQDYLGKIHGAYSSEKLSYFEHNLETWRQLWRVLEMSDIVLLITDIRHPVVNFPPALYEspecificaciones
NP_005266 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ELLELDIREVYQPGSVLDFPRRPPWSYEMSKEQLMSQEERSFQDYLGKIHGAYSSEKLSYFEHNLETWRQLWRVLEMSDIVLLITDIRHPVVNFPPALY | |
RUO | |
GNL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2794 | |
GNL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSR1 | |
GNL1 | |
Recombinant | |
wheat germ expression system |