missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GNG5 Partial ORF (NP_005265, 1 a.a. - 64 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002787-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSGSSSVAAMKKVVQQLRLEAGLNRVKVSQAAADLKQFCLQNAQHDPLLTGVSSSTNPFRPQKVEspecificaciones
NP_005265 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.78kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSGSSSVAAMKKVVQQLRLEAGLNRVKVSQAAADLKQFCLQNAQHDPLLTGVSSSTNPFRPQKV | |
RUO | |
GNG5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2787 | |
GNG5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ92393 | |
GNG5 | |
Recombinant | |
wheat germ expression system |