Learn More
Abnova™ Human GJB6 Partial ORF (AAH38934, 45 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010804-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Gap junctions allow the transport of ions and metabolites between the cytoplasm of adjacent cells. They are formed by two hemichannels, made up of six connexin proteins assembled in groups. Each connexin protein has four transmembrane segments, two extracellular loops, a cytoplasmic loop formed between the two inner transmembrane segments, and the N- and C-terminus both being in the cytoplasm. The specificity of the gap junction is determined by which connexin proteins comprise the hemichannel. In the past, connexin protein names were based on their molecular weight, however the new nomenclature uses sequential numbers based on which form (alpha or beta) of the gap junction is present. This gene encodes one of the connexin proteins. Mutations in this gene have been found in some forms of deafness and in some families with hidrotic ectodermal dysplasia. [provided by RefSeq]
Sequence: GDEQEDFVCNTLQPGCKNVCYDHFFPVSHIREspecificaciones
AAH38934 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
29.15kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GDEQEDFVCNTLQPGCKNVCYDHFFPVSHIR | |
RUO | |
GJB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10804 | |
GJB6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CX30/DFNA3/ED2/EDH/HED | |
GJB6 | |
Recombinant | |
wheat germ expression system |