Learn More
Abnova™ Human GDI2 Partial ORF (NP_001485, 346 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002665-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: QGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGEDEspecificaciones
NP_001485 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGED | |
RUO | |
GDI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2665 | |
GDI2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16452/FLJ37352/RABGDIB | |
GDI2 | |
Recombinant | |
wheat germ expression system |