Learn More
Abnova™ Human GDF7 Partial ORF (NP_878248, 361 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00151449-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord. [provided by RefSeq]
Sequence: LGWDDWIIAPLDYEAYHCEGLCDFPLRSHLEPTNHAIIQTLLNSMAPDAAPASCCVPARLSPISILYIDAANNVVYKQYEDMVVEACGCREspecificaciones
NP_878248 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGWDDWIIAPLDYEAYHCEGLCDFPLRSHLEPTNHAIIQTLLNSMAPDAAPASCCVPARLSPISILYIDAANNVVYKQYEDMVVEACGCR | |
RUO | |
GDF7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
151449 | |
GDF7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP12 | |
GDF7 | |
Recombinant | |
wheat germ expression system |