Learn More
Abnova™ Human FUT1 Full-length ORF (NP_000139.1, 1 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002523-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway. This gene is one of two encoding the galactoside 2-L-fucosyltransferase enzyme. Mutations in this gene are a cause of the H-Bombay blood group. [provided by RefSeq]
Sequence: MWLRSHRQLCLAFLLVCVLSVIFFLHIHQDSFPHGLGLSILCPDRRLVTPPVAIFCLPGTAMGPNASSSCPQHPASLSGTWTVYPNGRFGNQMGQYATLLALAQLNGRRAFILPAMHAALAPVFRITLPVLAPEVDSRTPWRELQLHDWMSEEYADLRDPFLKLSGFPCSWTFFHHLREQIRREFTLHDHLREEAQSVLGQLRLGRTGDRPRTFVGVHVRRGDYLQVMPQRWKGVVGDSAYLRQAMDWFRARHEAPVFVVTSNGMEWCKENIDTSQGDVTFAGDGQEATPWKDFALLTQCNHTIMTIGTFGFWAAYLAGGDTVYLANFTLPDSEFLKIFKPEAAFLPEWVGINADLSPLWTLAKPEspecificaciones
NP_000139.1 | |
Liquid | |
2523 | |
FUT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWLRSHRQLCLAFLLVCVLSVIFFLHIHQDSFPHGLGLSILCPDRRLVTPPVAIFCLPGTAMGPNASSSCPQHPASLSGTWTVYPNGRFGNQMGQYATLLALAQLNGRRAFILPAMHAALAPVFRITLPVLAPEVDSRTPWRELQLHDWMSEEYADLRDPFLKLSGFPCSWTFFHHLREQIRREFTLHDHLREEAQSVLGQLRLGRTGDRPRTFVGVHVRRGDYLQVMPQRWKGVVGDSAYLRQAMDWFRARHEAPVFVVTSNGMEWCKENIDTSQGDVTFAGDGQEATPWKDFALLTQCNHTIMTIGTFGFWAAYLAGGDTVYLANFTLPDSEFLKIFKPEAAFLPEWVGINADLSPLWTLAKP | |
RUO | |
FUT1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
H/HH/HSC | |
FUT1 | |
Yes | |
wheat germ expression system |