missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FPR3 Partial ORF (NP_002021.3, 254 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002359-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: WFPYELIGILMAVWLKEMLLNGKYKIILVLINPTSSLAFFNSCLNPILYVFMGRNFQERLIRSLPTSLERALTEVPDSAQTSNTDTTSASPPEETELQAMEspecificaciones
NP_002021.3 | |
Liquid | |
2359 | |
FPR3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FML2_HUMAN/FMLPY/FPRH1/FPRH2/FPRL2/RMLP-R-I | |
FPR3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
WFPYELIGILMAVWLKEMLLNGKYKIILVLINPTSSLAFFNSCLNPILYVFMGRNFQERLIRSLPTSLERALTEVPDSAQTSNTDTTSASPPEETELQAM | |
RUO | |
FPR3 | |
Wheat Germ (in vitro) | |
GST |