Learn More
Abnova™ Human FOXI1 Full-length ORF (NP_658982.1, 1 a.a. - 283 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002299-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSSFDLPAPSPPRCSPQFPSIGQEPPEMNLYYENFFHPQGVPSPQRPSFEGGGEYGATPNPYLWFNGPTMTPPPYLPGPNASPFLPQAYGVQRPLLPSVSGLGGSDLGWLPIPSQEELMKLVRPPYSYSALIAMAIHGAPDKRLTLSQIYQYVADNFPFYNKSKAGWQNSIRHNLSLNDCFKKVPRDEDDPAYVSGGSPTSHPLVTPGLSPEPSDKTGQNSLTFNSFSPLTNLSNHSGGGDWANPMPTNMLSYGGSVLSQFSPHFYNSVNTSGVLYPREGTEVEspecificaciones
NP_658982.1 | |
Liquid | |
2299 | |
FOXI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSFDLPAPSPPRCSPQFPSIGQEPPEMNLYYENFFHPQGVPSPQRPSFEGGGEYGATPNPYLWFNGPTMTPPPYLPGPNASPFLPQAYGVQRPLLPSVSGLGGSDLGWLPIPSQEELMKLVRPPYSYSALIAMAIHGAPDKRLTLSQIYQYVADNFPFYNKSKAGWQNSIRHNLSLNDCFKKVPRDEDDPAYVSGGSPTSHPLVTPGLSPEPSDKTGQNSLTFNSFSPLTNLSNHSGGGDWANPMPTNMLSYGGSVLSQFSPHFYNSVNTSGVLYPREGTEV | |
RUO | |
FOXI1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.20kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKH10/FKHL10/FREAC6/HFH3/MGC34197 | |
FOXI1 | |
Yes | |
wheat germ expression system |