missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXA1 Full-length ORF (NP_004487.2, 1 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003169-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198827

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. [provided by RefSeq]

Sequence: MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYANPGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSPMAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQNQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGSGSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELKTPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAYEQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTS
Especificaciones

Especificaciones

NP_004487.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HNF3A/MGC33105/TCF3A
FOXA1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3169
FOXA1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYANPGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSPMAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQNQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGSGSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELKTPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAYEQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTS
RUO
FOXA1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales