Learn More
Invitrogen™ Human Follistatin-like 1 ELISA Kit
Human Follistatin-like 1 quantitates human Follistatin-like 1 in serum, plasma, supernatant.
Marca: Invitrogen™ EH198RBX10
Detalles adicionales : Peso : 2.50000kg
Descripción
Human Follistatin-like 1 quantitates human Follistatin-like 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Follistatin-like 1.
This gene encodes a protein with similarity to follistatin, an activin-binding protein. It contains an FS module, a follistatin-like sequence containing 10 conserved cysteine residues. This gene product is thought to be an autoantigen associated with rheumatoid arthritis.Especificaciones
AI316791; AW107808; FLJ50214; FLJ52277; follistatin like 1; follistatin-like 1; follistatin-like protein 1; Follistatin-related protein 1; FRP; FSL1; Fstl; FSTL1; MIR198; OCC1; OCC-1; TGF-beta-inducible protein TSC-36; tsc36; TSC-36 | |
0.55 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Fstl1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.55 to 125 ng/mL | |
Biotin | |
Supernatant | |
Q12841 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
FSTL1 | |
4 hr 45 min |