missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLJ25680 Partial ORF (NP_694948.1, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00134187-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: AGHRPSNHFCPLPGSGGGGPRGPMPLRVDTLTWLSTQAAPGRVMVWPAVRPGICPGPDVWRIPLGPLPHEFRGWIAPCRPRLGASEAGDWLRRPSEGALEspecificaciones
NP_694948.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AGHRPSNHFCPLPGSGGGGPRGPMPLRVDTLTWLSTQAAPGRVMVWPAVRPGICPGPDVWRIPLGPLPHEFRGWIAPCRPRLGASEAGDWLRRPSEGAL | |
RUO | |
POU5F2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
134187 | |
FLJ25680 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686P02123/FLJ25680/SPRM-1 | |
POU5F2 | |
Recombinant | |
wheat germ expression system |