Learn More
Abnova™ Human FGL2 Full-length ORF (NP_006673.1, 1 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010875-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a secreted protein that is similar to the beta- and gamma-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. This protein may play a role in physiologic functions at mucosal sites. [provided by RefSeq]
Sequence: MKLANWYWLSSAVLATYGFLVVANNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPGEVGDNRVRELESEVNKLSSELKNAKEEINVLHGRLEKLNLVNMNNIENYVDSKVANLTFVVNSLDGKCSKCPSQEQIQSRPVQHLIYKDCSDYYAIGKRSSETYRVTPDPKNSSFEVYCDMETMGGGWTVLQARLDGSTNFTRTWQDYKAGFGNLRREFWLGNDKIHLLTKSKEMILRIDLEDFNGVELYALYDQFYVANEFLKYRLHVGNYNGTAGDALRFNKHYNHDLKFFTTPDKDNDRYPSGNCGLYYSSGWWFDACLSANLNGKYYHQKYRGVRNGIFWGTWPGVSEAHPGGYKSSFKEAKMMIRPKHFKPEspecificaciones
NP_006673.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
T49/pT49 | |
FGL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10875 | |
FGL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLANWYWLSSAVLATYGFLVVANNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPGEVGDNRVRELESEVNKLSSELKNAKEEINVLHGRLEKLNLVNMNNIENYVDSKVANLTFVVNSLDGKCSKCPSQEQIQSRPVQHLIYKDCSDYYAIGKRSSETYRVTPDPKNSSFEVYCDMETMGGGWTVLQARLDGSTNFTRTWQDYKAGFGNLRREFWLGNDKIHLLTKSKEMILRIDLEDFNGVELYALYDQFYVANEFLKYRLHVGNYNGTAGDALRFNKHYNHDLKFFTTPDKDNDRYPSGNCGLYYSSGWWFDACLSANLNGKYYHQKYRGVRNGIFWGTWPGVSEAHPGGYKSSFKEAKMMIRPKHFKP | |
RUO | |
FGL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |