missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO7 Partial ORF (NP_036311, 357 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00025793-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16117742

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined. [provided by RefSeq]

Sequence: AVCRDLFTASNDPLLWRFLYLRDFRDNTVRVQDTDWKELYRKRHIQRKESPKGRFVMLLPSSTHTIPFYPNPLHPRPFPSSRLPPGIIGGEYDQRPTLP
Especificaciones

Especificaciones

NP_036311
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AVCRDLFTASNDPLLWRFLYLRDFRDNTVRVQDTDWKELYRKRHIQRKESPKGRFVMLLPSSTHTIPFYPNPLHPRPFPSSRLPPGIIGGEYDQRPTLP
RUO
FBXO7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25793
FBXO7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686B08113/FBX/FBX07/FBX7/PARK15/PKPS
FBXO7
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales