Learn More
Abnova™ Human FBXL7 Full-length ORF (NP_036436.1, 1 a.a. - 491 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023194-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats. [provided by RefSeq]
Sequence: MGANNGKQYGSEGKGSSSISSDVSSSTDHTPTKAQKNVATSEDSDLSMRTLSTPSPALICPPNLPGFQNGRGSSTSSSSITGETVAMVHSPPPTRLTHPLIRLASRPQKEQASIDRLPDHSMVQIFSFLPTNQLCRCARVCRRWYNLAWDPRLWRTIRLTGETINVDRALKVLTRRLCQDTPNVCLMLETVTVSGCRRLTDRGLYTIAQCCPELRRLEVSGCYNISNEAVFDVVSLCPNLEHLDVSGCSKVTCISLTREASIKLSPLHGKQISIRYLDMTDCFVLEDEGLHTIAAHCTQLTHLYLRRCVRLTDEGLRYLVIYCASIKELSVSDCRFVSDFGLREIAKLESRLRYLSIAHCGRVTDVGIRYVAKYCSKLRYLNARGCEGITDHGVEYLAKNCTKLKSLDIGKCPLVSDTGLECLALNCFNLKRLSLKSCESITGQGLQIVAANCFDLQTLNVQDCEVSVEALRFVKRHCKRCVIEHTNPAFFEspecificaciones
NP_036436.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL6/FBL7 | |
FBXL7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23194 | |
FBXL7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGANNGKQYGSEGKGSSSISSDVSSSTDHTPTKAQKNVATSEDSDLSMRTLSTPSPALICPPNLPGFQNGRGSSTSSSSITGETVAMVHSPPPTRLTHPLIRLASRPQKEQASIDRLPDHSMVQIFSFLPTNQLCRCARVCRRWYNLAWDPRLWRTIRLTGETINVDRALKVLTRRLCQDTPNVCLMLETVTVSGCRRLTDRGLYTIAQCCPELRRLEVSGCYNISNEAVFDVVSLCPNLEHLDVSGCSKVTCISLTREASIKLSPLHGKQISIRYLDMTDCFVLEDEGLHTIAAHCTQLTHLYLRRCVRLTDEGLRYLVIYCASIKELSVSDCRFVSDFGLREIAKLESRLRYLSIAHCGRVTDVGIRYVAKYCSKLRYLNARGCEGITDHGVEYLAKNCTKLKSLDIGKCPLVSDTGLECLALNCFNLKRLSLKSCESITGQGLQIVAANCFDLQTLNVQDCEVSVEALRFVKRHCKRCVIEHTNPAFF | |
RUO | |
FBXL7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |