missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXL16 Full-length ORF (NP_699181.1, 1 a.a. - 479 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00146330-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198413

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Members of the F-box protein family, such as FBXL16, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MSSPGIDGDPKPPCLPRNGLVKLPGQPNGLGAASITKGTPATKNRPCQPPPPPTLPPPSLAAPLSRAALAGGPCTPAGGPASALAPGHPAERPPLATDEKILNGLFWYFSACEKCVLAQVCKAWRRVLYQPKFWAGLTPVLHAKELYNVLPGGEKEFVNLQGFAARGFEGFCLVGVSDLDICEFIDNYALSKKGVKAMSLKRSTITDAGLEVMLEQMQGVVRLELSGCNDFTEAGLWSSLSARITSLSVSDCINVADDAIAAISQLLPNLAELSLQAYHVTDTALAYFTARQGHSTHTLRLLSCWEITNHGVVNVVHSLPNLTALSLSGCSKVTDDGVELVAENLRKLRSLDLSWCPRITDMALEYVACDLHRLEELVLDRCVRITDTGLSYLSTMSSLRSLYLRWCCQVQDFGLKHLLALGSLRLLSPAGCPLLTTTGLSGLVQLQELEELELTNCPGATPELFKYFSQHLPRCLVIE
Especificaciones

Especificaciones

NP_699181.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C16orf22/FLJ33735/Fbl16/MGC33974/c380A1.1
FBXL16
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
146330
FBXL16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSPGIDGDPKPPCLPRNGLVKLPGQPNGLGAASITKGTPATKNRPCQPPPPPTLPPPSLAAPLSRAALAGGPCTPAGGPASALAPGHPAERPPLATDEKILNGLFWYFSACEKCVLAQVCKAWRRVLYQPKFWAGLTPVLHAKELYNVLPGGEKEFVNLQGFAARGFEGFCLVGVSDLDICEFIDNYALSKKGVKAMSLKRSTITDAGLEVMLEQMQGVVRLELSGCNDFTEAGLWSSLSARITSLSVSDCINVADDAIAAISQLLPNLAELSLQAYHVTDTALAYFTARQGHSTHTLRLLSCWEITNHGVVNVVHSLPNLTALSLSGCSKVTDDGVELVAENLRKLRSLDLSWCPRITDMALEYVACDLHRLEELVLDRCVRITDTGLSYLSTMSSLRSLYLRWCCQVQDFGLKHLLALGSLRLLSPAGCPLLTTTGLSGLVQLQELEELELTNCPGATPELFKYFSQHLPRCLVIE
RUO
FBXL16
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales