missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM100A Full-length ORF (NP_660296.1, 1 a.a. - 177 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00124402-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Description
Sequence: MSVNMDELKHQVMINQFVLTAGCAADQAKQLLQAAHWQFETALSAFFQETNIPYSHHHHQMMCTPANTPATPPNFPDALTMFSRLKASESFHSGGSGSPMAATATSPPPHFPHAATSSSAASSWPTAASPPGGPQHHQPQPPLWTPTPPSPASDWPPLAPQQATSEPRAHPAMEAERSpecifications
NP_660296.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31223/FLJ32185/PP11303 | |
FAM100A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
124402 | |
FAM100A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVNMDELKHQVMINQFVLTAGCAADQAKQLLQAAHWQFETALSAFFQETNIPYSHHHHQMMCTPANTPATPPNFPDALTMFSRLKASESFHSGGSGSPMAATATSPPPHFPHAATSSSAASSWPTAASPPGGPQHHQPQPPLWTPTPPSPASDWPPLAPQQATSEPRAHPAMEAER | |
RUO | |
FAM100A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |