Learn More
ACROBiosystems Human FABP5 / E-FABP Protein, His Tag
Marca: ACROBiosystems FA5-H5140-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Fatty acid-binding protein 5 (FABP5), is also known as Fatty acid-binding protein, epidermal (E-FABP), Psoriasis-associated fatty acid-binding protein homolog (PA-FABP). FABP5 / E-FABP belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. FABP5 / E-FABP is highly expressed in psoriatic skin. FABP5 / E-FABP has high specificity for fatty acids and has highest affinity for C18 chain length. FABP5 may be involved in keratinocyte differentiation.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.0 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).Especificaciones
AAH19385 | |
100 μg | |
Ala 2 - Glu 135 | |
FABP5,E-FABP,PA-FABP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
16.0 kDa | |
Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession # AAH19385). | |
-20°C | |
FABP5 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |