Learn More
Abnova™ Human ERN1 Full-length ORF (NP_689674.2, 1 a.a. - 70 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002081-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is the ER to nucleus signalling 1 protein, a human homologue of the yeast Ire1 gene product. This protein possesses intrinsic kinase activity and an endoribonuclease activity and it is important in altering gene expression as a response to endoplasmic reticulum-based stress signals. [provided by RefSeq]
Sequence: MPARRLLLLLTLLLPGLGVSDRGAWGGGQLATAGSGPGQRRGAGAGVRAGSATAAARCPVSPAVGGSGRASpecifica
NP_689674.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30999/IRE1/IRE1P/MGC163277/MGC163279 | |
ERN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2081 | |
ERN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPARRLLLLLTLLLPGLGVSDRGAWGGGQLATAGSGPGQRRGAGAGVRAGSATAAARCPVSPAVGGSGRA | |
RUO | |
ERN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |